Learn More
Tube AL-23, ouvert aux deux extrémités, dia. ext. : 8 mm, dia. int. de : 5mm, Thermo Scientific™
28.80€ - 129.00€
Spécification
Nom chimique ou matériau | Al-23 Tube |
---|---|
Note relative au nom | Both ends open |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11333947
|
Thermo Scientific Alfa Aesar
031560.DM |
100 mm | |||||||
11303947
|
Thermo Scientific Alfa Aesar
031560.DR |
200 mm | |||||||
11343947
|
Thermo Scientific Alfa Aesar
031560.DW |
300 mm | |||||||
11323947
|
Thermo Scientific Alfa Aesar
031560.EH |
600 mm | |||||||
11353947
|
Thermo Scientific Alfa Aesar
031560.EM |
800 mm | |||||||
11313947
|
Thermo Scientific Alfa Aesar
031560.EO |
1000 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Tube | |
Both ends open |
Sécurité et traitement
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures