Learn More
Tube isolant multibore Al-23, dia. ext. : 6mm, deux alésages de 1,5 mm, Thermo Scientific™
23.51€ - 239.00€
Spécification
Nom chimique ou matériau | Al-23 Multibore Insulating Tube |
---|---|
Note relative au nom | Two bores |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11358437
|
Thermo Scientific Alfa Aesar
032556.DS |
205 mm | |||||||
11378437
|
Thermo Scientific Alfa Aesar
032556.DV |
275 mm | |||||||
11318447
|
Thermo Scientific Alfa Aesar
032556.DZ |
380 mm | |||||||
11308447
|
Thermo Scientific Alfa Aesar
032556.EG |
560 mm | |||||||
11348437
|
Thermo Scientific Alfa Aesar
032556.EL |
770 mm | |||||||
11368437
|
Thermo Scientific Alfa Aesar
032556.ER |
1060 mm | |||||||
11398437
|
Thermo Scientific Alfa Aesar
032556.EW |
1460 mm | |||||||
11388437
|
Thermo Scientific Alfa Aesar
032556.FB |
2060 mm | |||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Al-23 Multibore Insulating Tube | |
Two bores |
Sécurité et traitement
missing translation for 'tsca' : U
Recommended Storage : Ambient temperatures