missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acide 4-fluoronaphtalène-1-boronique, 98 %, Thermo Scientific™
47.75€ - 136.00€
Identifiants chimiques
CAS | 182344-25-8 |
---|---|
Formule moléculaire | C10H8BFO2 |
Poids moléculaire (g/mol) | 189.98 |
Numéro MDL | MFCD04039221 |
Clé InChI | IDCGAEJGZLYEAZ-UHFFFAOYSA-N |
Synonyme | 4-fluoronaphthalene-1-boronic acid, 4-fluoronaphtalene-1-boronic acid, 4-fluoronaphthalen-1-yl-1-boronic acid, 4-fluoronaphthalen-1-yl boronic acid, 4-fluoro-1-naphthyl boronic acid, 4-fluoro-1-naphthaleneboronic acid, boronic acid, 4-fluoro-1-naphthalenyl, akos brn-0210, acmc-209ejb, 4-fluoronaphth-1-ylboronic acid |
CID PubChem | 2782675 |
Nom IUPAC | Acide (4-fluoronaphthalén-1-yl)boronique |
SMILES | B(C1=CC=C(C2=CC=CC=C12)F)(O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
182344-25-8 | |
189.98 | |
IDCGAEJGZLYEAZ-UHFFFAOYSA-N | |
2782675 | |
B(C1=CC=C(C2=CC=CC=C12)F)(O)O |
C10H8BFO2 | |
MFCD04039221 | |
4-fluoronaphthalene-1-boronic acid, 4-fluoronaphtalene-1-boronic acid, 4-fluoronaphthalen-1-yl-1-boronic acid, 4-fluoronaphthalen-1-yl boronic acid, 4-fluoro-1-naphthyl boronic acid, 4-fluoro-1-naphthaleneboronic acid, boronic acid, 4-fluoro-1-naphthalenyl, akos brn-0210, acmc-209ejb, 4-fluoronaphth-1-ylboronic acid | |
Acide (4-fluoronaphthalén-1-yl)boronique |
Spécification
4-Fluoronaphthalene-1-boronic acid | |
240°C to 242°C | |
MFCD04039221 | |
IDCGAEJGZLYEAZ-UHFFFAOYSA-N | |
Acide (4-fluoronaphthalén-1-yl)boronique | |
2782675 | |
98% |
182344-25-8 | |
C10H8BFO2 | |
4-fluoronaphthalene-1-boronic acid, 4-fluoronaphtalene-1-boronic acid, 4-fluoronaphthalen-1-yl-1-boronic acid, 4-fluoronaphthalen-1-yl boronic acid, 4-fluoro-1-naphthyl boronic acid, 4-fluoro-1-naphthaleneboronic acid, boronic acid, 4-fluoro-1-naphthalenyl, akos brn-0210, acmc-209ejb, 4-fluoronaphth-1-ylboronic acid | |
B(C1=CC=C(C2=CC=CC=C12)F)(O)O | |
189.98 | |
189.98 |
Sécurité et traitement
Déclaration GHS H
H315-H319-H335
Provoque une irritation cutanée.
Provoque une grave irritation oculaire.
Peut irriter les voies respiratoires.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : Non
Recommended Storage : Température ambiante