missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Octanoylthiophène, 99 %, Thermo Scientific™
21.30€ - 274.00€
Identifiants chimiques
CAS | 30711-41-2 |
---|---|
Formule moléculaire | C12H18OS |
Poids moléculaire (g/mol) | 210.335 |
Numéro MDL | MFCD00053146 |
Clé InChI | QRJZCQOLJVIBDE-UHFFFAOYSA-N |
Synonyme | 2-octanoylthiophene, 2-n-octanoylthiophene, 2-octanoyl thiophene, n-heptyl 2-thienyl ketone, 1-thiophen-2-yl octan-1-one, 1-octanone, 1-2-thienyl, thiophene, 2-octanoyl, acmc-1ad13, 1-2-thienyl octan-1-one |
CID PubChem | 141582 |
Nom IUPAC | 1-thiophène-2-yloctan-1-one |
SMILES | CCCCCCCC(=O)C1=CC=CS1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
30711-41-2 | |
210.335 | |
QRJZCQOLJVIBDE-UHFFFAOYSA-N | |
141582 | |
CCCCCCCC(=O)C1=CC=CS1 |
C12H18OS | |
MFCD00053146 | |
2-octanoylthiophene, 2-n-octanoylthiophene, 2-octanoyl thiophene, n-heptyl 2-thienyl ketone, 1-thiophen-2-yl octan-1-one, 1-octanone, 1-2-thienyl, thiophene, 2-octanoyl, acmc-1ad13, 1-2-thienyl octan-1-one | |
1-thiophène-2-yloctan-1-one |
Spécification
2-Octanoylthiophene | |
1.008 | |
C12H18OS | |
MFCD00053146 | |
2-octanoylthiophene, 2-n-octanoylthiophene, 2-octanoyl thiophene, n-heptyl 2-thienyl ketone, 1-thiophen-2-yl octan-1-one, 1-octanone, 1-2-thienyl, thiophene, 2-octanoyl, acmc-1ad13, 1-2-thienyl octan-1-one | |
CCCCCCCC(=O)C1=CC=CS1 | |
210.335 | |
210.34 |
30711-41-2 | |
120°C to 122°C (1 mmHg) | |
1.52 | |
4220 | |
QRJZCQOLJVIBDE-UHFFFAOYSA-N | |
1-thiophène-2-yloctan-1-one | |
141582 | |
99% |
Sécurité et traitement
missing translation for 'tsca' : No
Recommended Storage : Température ambiante