Learn More
2’,4’-Difluoro-3’-méthoxyacétophénone, 97 %
83.28€ - 275.00€
Identifiants chimiques
CAS | 373603-19-1 |
---|---|
Formule moléculaire | C9H8F2O2 |
Poids moléculaire (g/mol) | 186.158 |
Numéro MDL | MFCD02258906 |
Clé InChI | KTZOZLGEZVDGMS-UHFFFAOYSA-N |
Synonyme | 2',4'-difluoro-3'-methoxyacetophenone, 2,4-difluoro-3-methoxy acetophenone, 2,4-difluoro-3-methoxyacetophenone, 1-2,4-difluoro-3-methoxyphenyl ethanone, 1-2,4-difluoro-3-methoxy-phenyl ethanone, 1-acetyl-2,4-difluoro-3-methoxybenzene, 1-2,4-difluoro-3-methoxyphenyl ethan-1-one |
CID PubChem | 3850760 |
Nom IUPAC | 1-(2,4-difluoro-3-méthoxyphényl)éthanone |
SMILES | CC(=O)C1=C(C(=C(C=C1)F)OC)F |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identifiants chimiques
373603-19-1 | |
186.158 | |
KTZOZLGEZVDGMS-UHFFFAOYSA-N | |
3850760 | |
CC(=O)C1=C(C(=C(C=C1)F)OC)F |
C9H8F2O2 | |
MFCD02258906 | |
2',4'-difluoro-3'-methoxyacetophenone, 2,4-difluoro-3-methoxy acetophenone, 2,4-difluoro-3-methoxyacetophenone, 1-2,4-difluoro-3-methoxyphenyl ethanone, 1-2,4-difluoro-3-methoxy-phenyl ethanone, 1-acetyl-2,4-difluoro-3-methoxybenzene, 1-2,4-difluoro-3-methoxyphenyl ethan-1-one | |
1-(2,4-difluoro-3-méthoxyphényl)éthanone |
Spécification
2',4'-Difluoro-3'-methoxyacetophenone | |
C9H8F2O2 | |
MFCD02258906 | |
KTZOZLGEZVDGMS-UHFFFAOYSA-N | |
1-(2,4-difluoro-3-méthoxyphényl)éthanone | |
3850760 | |
97% |
373603-19-1 | |
1.494 | |
2',4'-difluoro-3'-methoxyacetophenone, 2,4-difluoro-3-methoxy acetophenone, 2,4-difluoro-3-methoxyacetophenone, 1-2,4-difluoro-3-methoxyphenyl ethanone, 1-2,4-difluoro-3-methoxy-phenyl ethanone, 1-acetyl-2,4-difluoro-3-methoxybenzene, 1-2,4-difluoro-3-methoxyphenyl ethan-1-one | |
CC(=O)C1=C(C(=C(C=C1)F)OC)F | |
186.158 | |
186.15 |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures