missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 sigma Mouse anti-Human, Clone: 3F10D9, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 66251-1-IG-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a cell cycle checkpoint protein. The encoded protein binds to translation and initiation factors and functions as a regulator of mitotic translation. In response to DNA damage this protein plays a role in preventing DNA errors during mitosis.Spécification
14-3-3 sigma | |
Monoclonal | |
1.37 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P31947 | |
SFN | |
14-3-3 sigma Fusion Protein Ag12644 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
3F10D9 | |
Unconjugated | |
SFN | |
14 3 3 protein sigma, HME1, SFN, stratifin, YWHAS | |
Mouse | |
Protein G | |
RUO | |
2810 | |
-20°C | |
Liquid |