missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ 1-tert-Butyl-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde, 95%
138.00€ - 622.00€
Identifiants chimiques
CAS | 647824-51-9 |
---|---|
Formule moléculaire | C10H16N2O |
Poids moléculaire (g/mol) | 180.251 |
Numéro MDL | MFCD00120293 |
Clé InChI | WQHXOHHEPNBXMF-UHFFFAOYSA-N |
Synonyme | 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c |
CID PubChem | 2776936 |
Nom IUPAC | 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde |
SMILES | CC1=C(C(=NN1C(C)(C)C)C)C=O |
Description
Identifiants chimiques
647824-51-9 | |
180.251 | |
WQHXOHHEPNBXMF-UHFFFAOYSA-N | |
2776936 | |
CC1=C(C(=NN1C(C)(C)C)C)C=O |
C10H16N2O | |
MFCD00120293 | |
1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c | |
1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde |
Spécification
1-tert-Butyl-3,5-dimethyl-1H-pyrazole-4-carboxaldehyde | |
C10H16N2O | |
1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carbaldehyde, 1-tert-butyl-3,5-dimethyl-1h-pyrazole-4-carboxaldehyde, buttpark 99\12-15, 1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde, acmc-20ap9c | |
CC1=C(C(=NN1C(C)(C)C)C)C=O | |
180.251 | |
180.25 |
647824-51-9 | |
MFCD00120293 | |
WQHXOHHEPNBXMF-UHFFFAOYSA-N | |
1-tert-butyl-3,5-dimethylpyrazole-4-carbaldehyde | |
2776936 | |
95% |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning