Learn More
Cole Parmer Syringe pump lock tip glass 230V 50Hz 1mL graduations 20mL Masterflex, 1/Cs, MA: TBD
Marque:  Cole Parmer 25701-51
Syringe pump lock tip glass 230V 50Hz 1mL graduations 20mL Masterflex, 1/Cs, MA: TBD
Code nomenclature Nacres: NB.03
Informations supplémentaires : Poids : 0.01000kg
Code produit. 12486250