Learn More
Thermo Scientific Alfa Aesar Aluminum nitride crucible, Cylindrical, Flat Base,Height x OD x ID (mm), 37 x 33.5 x 28.7, Vol(ml),
Marque:  Thermo Scientific Alfa Aesar 043350.KF
Aluminum nitride crucible, Cylindrical, Flat Base,Height x OD x ID (mm), 37 x 33.5 x 28.7, Vol(ml),
Code nomenclature Nacres: NB.45
Informations supplémentaires : Poids : 0.01000kg
Code produit. 15423895