Learn More
ZNF366 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24340-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The ZNF366 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ZNF366. This antibody recognizes human, mouse antigen. The ZNF366 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
ZNF366 | |
Polyclonal | |
Unconjugated | |
20 μL | |
167465 | |
IgG |
Western Blot, ELISA | |
420 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse |