Learn More
ZNF366 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 24340-1-AP-150UL
Code produit. 16812825
  • 399.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
The ZNF366 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human ZNF366. This antibody recognizes human, mouse antigen. The ZNF366 antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
Spécification
ZNF366
Polyclonal
Unconjugated
150 μL
167465
IgG
Western Blot, ELISA
420 μg/mL
Rabbit
Primary
Human, Mouse