Learn More
ZDHHC15 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 21627-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene belongs to the DHHC palmitoyltransferase family. Mutations in this gene are associated with mental retardation X-linked type 91 (MRX91). Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spécification
ZDHHC15 | |
Polyclonal | |
Unconjugated | |
ZDHHC15 | |
DHHC 15, MRX91, Palmitoyltransferase ZDHHC15, ZDHHC15 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
158866 | |
-20°C | |
Liquid |
Western Blot | |
0.15 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q96MV8 | |
ZDHHC15 | |
ZDHHC15 Fusion Protein Ag16152 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |