missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VPS54 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 13327-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes for a protein that in yeast forms part of a trimeric vacuolar-protein-sorting complex that is required for retrograde transport of proteins from prevacuoles to the late Golgi compartment. As in yeast, mammalian Vps54 proteins contain a coiled-coil region and dileucine motifs. Alternative splicing results in multiple transcript variants encoding different isoformsSpécification
VPS54 | |
Polyclonal | |
Unconjugated | |
VPS54 | |
HCC8, hVps54L, SLP 8p, Tumor antigen HOM HCC 8, Tumor antigen SLP 8p, VPS54, VPS54L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
245944, 286932, 51542 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q5SPW0, Q9JMK8, Q9P1Q0 | |
Vps54 | |
VPS54 Fusion Protein Ag4123 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |