missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRIM63 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 55456-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody is specific to TRIM63.
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin′s N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle.Spécification
TRIM63 | |
Polyclonal | |
Unconjugated | |
TRIM63 | |
IRF, Iris RING finger protein, MuRF 1, MURF1, MURF2, RING finger protein 28, RNF28, SMRZ, TRIM63, tripartite motif containing 63 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
140939, 433766, 84676 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence | |
0.67 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q38HM4, Q91Z63, Q969Q1 | |
Trim63 | |
Peptide | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |