Learn More
TRIM63 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 55456-1-AP-150UL
Code produit. 16866925
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
This antibody is specific to TRIM63.
This gene encodes a member of the RING zinc finger protein family found in striated muscle and iris. The product of this gene is localized to the Z-line and M-line lattices of myofibrils, where titin′s N-terminal and C-terminal regions respectively bind to the sarcomere. In vitro binding studies have shown that this protein also binds directly to titin near the region of titin containing kinase activity. Another member of this protein family binds to microtubules. Since these family members can form heterodimers, this suggests that these proteins may serve as a link between titin kinase and microtubule-dependent signal pathways in muscle.
Spécification
Spécification
TRIM63
Polyclonal
Unconjugated
TRIM63
IRF, Iris RING finger protein, MuRF 1, MURF1, MURF2, RING finger protein 28, RNF28, SMRZ, TRIM63, tripartite motif containing 63
Rabbit
Antigen Affinity Chromatography
RUO
140939, 433766, 84676
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.67 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q38HM4, Q91Z63, Q969Q1
Trim63
Peptide
150 μL
Primary
Human, Mouse, Rat