missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TRAIP Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | TRAIP |
---|---|
Concentration | 0.3 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16866653
|
Proteintech
10332-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16856653
|
Proteintech
10332-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
TRAIP is a protein that contains an N-terminal RING finger motif and a putative coiled-coil domain. A similar murine protein interacts with TNFR-associated factor 1 (TRAF1), TNFR-associated factor 2 (TRAF2), and cylindromatosis. The interaction with TRAF2 inhibits TRAF2-mediated nuclear factor kappa-B, subunit 1 activation that is required for cell activation and protection against apoptosis.Spécification
TRAIP | |
Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q8VIG6, Q9BWF2 | |
10293, 22036 | |
TRAIP Fusion Protein Ag0491 | |
Primary | |
-20°C | |
TRAIP |
0.3 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
RING finger protein 206, RNF206, TRAF interacting protein, TRAIP, TRIP | |
TRAIP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |