Learn More
Copan Italia™ X500 ECOUVILLON AMIES SANS CHARBON CHARBON
Single plastic swab suitable for throat, vaginal, wound, and skin swabs. Copan Italia™ Amies Agar Gel Swab, No Charcoal is coated with Amies agar gel, and designed for specimen transportation.
Marque: Copan Italia™ 108C.USE
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
- Single plastic swab
- Amies gel medium
- Suitable for throat, vaginal, wound, and skin swabs
- For specimen transportation
Spécification
Amies Agar | |
Throat, vaginal, wound, and skin swabs. For aerobes, anerobes, and fastidious bacteria culture, and gram staining. | |
50/Pk., 10 x 50/Cs. |
Blue | |
Plastic |