Learn More
STC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 20621-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a secreted, homodimeric glycoprotein that is expressed in a wide variety of tissues and may have autocrine or paracrine functions. The gene contains a 5′ UTR rich in CAG trinucleotide repeats. The encoded protein contains 11 conserved cysteine residues and is phosphorylated by protein kinase C exclusively on its serine residues. The protein may play a role in the regulation of renal and intestinal calcium and phosphate transport, cell metabolism, or cellular calcium/phosphate homeostasis. Overexpression of human stanniocalcin 1 in mice produces high serum phosphate levels, dwarfism, and increased metabolic rate. This gene has altered expression in hepatocellular, ovarian, and breast cancers.Especificaciones
STC1 | |
Polyclonal | |
Unconjugated | |
STC1 | |
stanniocalcin 1, STC, STC 1, STC1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20855, 6781, 81801 | |
-20°C | |
Liquid |
Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O55183, P52823, P97574 | |
STC1 | |
STC1 Fusion Protein Ag14697 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |