Learn More
SPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 12105-1-AP-20UL
Code produit. 16819623
  • 140.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
SPIN1 (spindlin 1), also known as SPIN or ovarian cancer-related protein (OCR), is a 262 amino acid nuclear protein suggested to play a role in regulation of the cell-cycle during the transition from gamete to embryo. A member of the SPIN/STSY family, SPIN1 localizes to interphase nucleus and mitotic chromosomes, and is modified by phosphorylation in a cell-cycle-dependent fashion. A meiotic spindle-binding protein, SPIN1 overexpression has been shown to cause defects in mitotic spindles, thereby resulting in chromosome instability and potential tumorigenesis.
Spécification
Spécification
SPIN1
Polyclonal
Unconjugated
SPIN1
OCR, Ovarian cancer related protein, SPIN, SPIN1, spindlin 1
Rabbit
Antigen Affinity Chromatography
RUO
10927, 20729, 361217
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunofluorescence, Immunocytochemistry, Immunoprecipitation
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q4V8J7, Q61142, Q9Y657
Spin1
SPIN1 Fusion Protein Ag2750
20 μL
Primary
Human, Rat, Mouse