missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPIN1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12105-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
SPIN1 (spindlin 1), also known as SPIN or ovarian cancer-related protein (OCR), is a 262 amino acid nuclear protein suggested to play a role in regulation of the cell-cycle during the transition from gamete to embryo. A member of the SPIN/STSY family, SPIN1 localizes to interphase nucleus and mitotic chromosomes, and is modified by phosphorylation in a cell-cycle-dependent fashion. A meiotic spindle-binding protein, SPIN1 overexpression has been shown to cause defects in mitotic spindles, thereby resulting in chromosome instability and potential tumorigenesis.Spécification
SPIN1 | |
Polyclonal | |
Unconjugated | |
SPIN1 | |
OCR, Ovarian cancer related protein, SPIN, SPIN1, spindlin 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10927, 20729, 361217 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4V8J7, Q61142, Q9Y657 | |
Spin1 | |
SPIN1 Fusion Protein Ag2750 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |