Learn More
Phosphate de sodium, solution tampon de 0,2M, pH 9,5, Thermo Scientific™
48.70€ - 63.60€
Spécification
Nom chimique ou matériau | Sodium phosphate buffer soln. |
---|---|
Concentration | Solution de tampon 0,2M |
pH | 9.5 |
Forme physique | Liquide |
Numéro MDL | MFCD00146206 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Spécification
Sodium phosphate buffer soln. | |
9.5 | |
MFCD00146206 |
Solution de tampon 0,2M | |
Liquide |
Sécurité et traitement
missing translation for 'tsca' : Oui
Recommended Storage : Température ambiante