missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMARCE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10814-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is part of the large ATP-dependent chromatin remodeling complex SWI/SNF, which is required for transcriptional activation of genes normally repressed by chromatin. The encoded protein, either alone or when in the SWI/SNF complex, can bind to 4-way junction DNA, which is thought to mimic the topology of DNA as it enters or exits the nucleosome. The protein contains a DNA-binding HMG domain, but disruption of this domain does not abolish the DNA-binding or nucleosome-displacement activities of the SWI/SNF complex. Unlike most of the SWI/SNF complex proteins, this protein has no yeast counterpart.Spécification
SMARCE1 | |
Polyclonal | |
Unconjugated | |
SMARCE1 | |
BAF57, BRG1 associated factor 57, SMARCE1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
303518, 57376, 6605 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.43 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O54941, Q969G3 | |
SMARCE1 | |
SMARCE1 Fusion Protein Ag1161 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |