missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC26A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 27759-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The SLC26 family is comprised of sulfate/anion transporters that are well conserved in both their genomic and protein structures, yet have markedly different tissue expression patterns. Members of the SLC26 family can mediate the electroneutral exchange of Cl- for HCO3- across the plasma membrane of mammalian cells. SLC26A2 (solute carrier family 26, member 2), also known as DTD, EDM4, DTDST, MST153, D5S1708 or MSTP157, is a 739 amino acid diastrophic dysplasia sulfate transporter belonging to the SLC26A/SulP transporter family. Ubiquitously expressed, SLC26A2 may play a role in endochondral bone formation. SLC26A2 is a transmembrane glycoprotein implicated in the pathogenesis of several human chondrodysplasias and is critical for sulfation of proteoglycans and matrix organization in cartilage.Spécification
SLC26A2 | |
Polyclonal | |
Unconjugated | |
SLC26A2 | |
D5S1708, Diastrophic dysplasia protein, DTD, DTDST, EDM4, MST153, MSTP157, SLC26A2, Sulfate transporter | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
117267, 13521, 1836 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O70531, P50443, Q62273 | |
SLC26A2 | |
SLC26A2 Fusion Protein Ag26521 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |