missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHPK Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12441-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene has weak homology to several carbohydrate kinases, a class of proteins involved in the phosphorylation of sugars as they enter a cell, inhibiting return across the cell membrane. Sequence variation between this novel gene and known carbohydrate kinases suggests the possibility of a different substrate, cofactor or changes in kinetic properties distinguishing it from other carbohydrate kinases. The gene resides in a region commonly deleted in cystinosis patients, suggesting a role as a modifier for the cystinosis phenotype. The genomic region is also rich in Alu repetitive sequences, frequently involved in chromosomal rearrangements.Spécification
SHPK | |
Polyclonal | |
Unconjugated | |
SHPK | |
CARKL, sedoheptulokinase, SHK, SHPK | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23729, 287479, 74637 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D5J6, Q9UHJ6 | |
Shpk | |
SHPK Fusion Protein Ag3150 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |