missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10054-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Signaling adapter that couples activated growth factor receptors to signaling pathways. Participates in signaling downstream of the angiopoietin receptor TEK/TIE2, and plays a role in the regulation of endothelial cell migration and sprouting angiogenesis (By similarity). Participates in a signaling cascade initiated by activated KIT and KITLG/SCF. Isoform p47Shc and isoform p52Shc, once phosphorylated, couple activated receptor kinases to Ras via the recruitment of the GRB2/SOS complex and are implicated in the cytoplasmic propagation of mitogenic signals. Isoform p47Shc and isoform p52 may thus function as initiators of the Ras signaling cascade in various non-neuronal systems. Isoform p66Shc does not mediate Ras activation, but is involved in signal transduction pathways that regulate the cellular response to oxidative stress and life span. Isoform p66Shc acts as a downstream target of the tumor suppressor p53 and is indispensable for the ability of stress-activated p53 to induce elevation of intracellular oxidants, cytochrome c release and apoptosis. The expression of isoform p66Shc has been correlated with life span.Spécification
SHC | |
Polyclonal | |
Unconjugated | |
SHC1 | |
SH2 domain protein C1, SHC, SHC transforming protein 1, SHC transforming protein 3, SHC transforming protein A, SHC1, SHCA | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20416, 6464 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
0.31 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P29353, P98083 | |
SHC1 | |
SHC Fusion Protein Ag0103 | |
20 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |