Learn More
Sestrin 2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10795-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a member of the sestrin family of PA26-related proteins. The encoded protein may function in the regulation of cell growth and survival. This protein may be involved in cellular response to different stress conditions.Spécification
Sestrin 2 | |
Polyclonal | |
Unconjugated | |
SESN2 | |
HI95, SES2, SESN2, SEST2, Sestrin2, Sestrin-2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
230784, 502988, 83667 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot, Immunoprecipitation | |
0.48 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P58004, P58043 | |
Sesn2 | |
Sestrin 2 Fusion Protein Ag1247 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |