missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SERPINE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11303-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.Spécification
SERPINE2 | |
Polyclonal | |
Unconjugated | |
SERPINE2 | |
GDN, Glia derived nexin, Peptidase inhibitor 7, PI 7, PI7, PN 1, PN1, PNI, Protease nexin 1, Protease nexin I, Serpin E2, SERPINE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20720, 29366, 5270 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunoprecipitation | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P07092, P07093, Q07235 | |
SERPINE2 | |
SERPINE2 Fusion Protein Ag1830 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |