Learn More
SERPINE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 11303-1-AP-150UL
Code produit. 16858283
  • 410.00€ / 150µL
Découvrez plus d'offres spéciales
Description
Description
This gene encodes a member of the serpin family of proteins, a group of proteins that inhibit serine proteases. Thrombin, urokinase, plasmin and trypsin are among the proteases that this family member can inhibit. This gene is a susceptibility gene for chronic obstructive pulmonary disease and for emphysema. Alternative splicing results in multiple transcript variants.
Spécification
Spécification
SERPINE2
Polyclonal
Unconjugated
SERPINE2
GDN, Glia derived nexin, Peptidase inhibitor 7, PI 7, PI7, PN 1, PN1, PNI, Protease nexin 1, Protease nexin I, Serpin E2, SERPINE2
Rabbit
Antigen Affinity Chromatography
RUO
20720, 29366, 5270
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P07092, P07093, Q07235
SERPINE2
SERPINE2 Fusion Protein Ag1830
150 μL
Primary
Human, Mouse, Rat