Learn More
Seladin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 10471-1-AP-20UL
 Afficher plus de versions de ce produit
Code produit. 16846873
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of its gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.
Spécification
Spécification
Seladin 1
Polyclonal
Unconjugated
DHCR24
DHCR24, Diminuto/dwarf1 homolog, KIAA0018, Nbla03646, Seladin 1, SELADIN1
Rabbit
Antigen Affinity Chromatography
RUO
1718, 298298, 74754
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.13 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q15392, Q5BQE6, Q8VCH6
DHCR24
Seladin 1 Fusion Protein Ag0756
20 μL
Primary
Rat, Human, Mouse