missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Seladin 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10471-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
DHCR24 is a flavin adenine dinucleotide (FAD)-dependent oxidoreductase which catalyzes the reduction of the delta-24 double bond of sterol intermediates during cholesterol biosynthesis. This protein contains a leader sequence that directs it to the endoplasmic reticulum membrane. Missense mutations in this gene have been associated with desmosterolosis. Also, reduced expression of its gene occurs in the temporal cortex of Alzheimer disease patients and overexpression has been observed in adrenal gland cancer cells.Spécification
Seladin 1 | |
Polyclonal | |
Unconjugated | |
DHCR24 | |
DHCR24, Diminuto/dwarf1 homolog, KIAA0018, Nbla03646, Seladin 1, SELADIN1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
1718, 298298, 74754 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q15392, Q5BQE6, Q8VCH6 | |
DHCR24 | |
Seladin 1 Fusion Protein Ag0756 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |