missing translation for 'onlineSavingsMsg'
Learn More

BUCHI Scrubber K-415 TripleScrub

A three-step gas scrubbing instrument with a powerful pump and condensation, neutralization and adsorption steps. It protects the user and the environment not only by neutralizing poisonous fumes and reaction gases, but also by minimizing the use of cooling water.

7047.32€

Spécification

Affichage None
Hertz 50/60 Hz
Puissance 140 W
Débit 32 LPM
À utiliser avec (application) Neutralizing Poisonous Fumes and Reaction Gases
Consulter d'autres spécifications

Produits 1
Code produit Marque Tension Dimensions (L x l x H) Type de produit Prix Quantité et disponibilité  
Code produit Marque Tension Dimensions (L x l x H) Type de produit Prix Quantité et disponibilité  
15202002
Afficher les documents Détails de la promotion
BUCHI
114152331
230 V 417 x 347 x 543 mm TripleScrubECO
7047.32€
1 pièce
Expédition estimée: 11-12-2024
pour voir le stock disponible 
Ajouter au panier
 
Description

Description

  • Aggressive acid or alkali fumes are sucked off by means of the powerful pump into the first stage for precondensation to avoid dilution of the second stage
  • The output from the condensation stage is neutralized in the neutralization vessel
  • Particles of organic and inorganic substances are collected in the third stage
  • Suction power can be adjusted anytime to allow for flexibility
  • K-415 can be controlled via a digester
  • For use with recirculation chillers
Spécification

Spécification

None
140 W
Neutralizing Poisonous Fumes and Reaction Gases
50/60 Hz
32 LPM
Suction: 50 to 400 mbar
Vidéos
FDS
Documents

Documents

Product Certifications