missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RRP7A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 24490-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
In cooperation with other chaperones, Hsp70s stabilize preexistent proteins against aggregation and mediate the folding of newly translated polypeptides in the cytosol as well as within organelles. These chaperones participate in all these processes through their ability to recognize nonnative conformations of other proteins. They bind extended peptide segments with a net hydrophobic character exposed by polypeptides during translation and membrane translocation, or following stress-induced damage.Especificaciones
RRP7A | |
Polyclonal | |
Unconjugated | |
RRP7A | |
BK126B4.3, CGI 96, Gastric cancer antigen Zg14, RRP7A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
27341, 74778 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
0.37 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9D1C9, Q9Y3A4 | |
Rrp7a | |
RRP7A Fusion Protein Ag21636 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |