Learn More
RPL23 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 16086-1-AP-20UL
 Afficher plus de versions de ce produit
Code produit. 16835574
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. It is present in protein complexes involved in histone acetylation and chromatin assembly. It is part of the Mi-2 complex which has been implicated in chromatin remodeling and transcriptional repression associated with histone deacetylation. This encoded protein is also part of co-repressor complexes, which is an integral component of transcriptional silencing. It is found among several cellular proteins that bind directly to retinoblastoma protein to regulate cell proliferation. This protein also seems to be involved in transcriptional repression of E2F-responsive genes. Three transcript variants encoding different isoforms have been found for this gene.
Spécification
Spécification
RPL23
Polyclonal
Unconjugated
RPL23
60S ribosomal protein L17, 60S ribosomal protein L23, ribosomal protein L23, rpL17, RPL23
Rabbit
Antigen Affinity Chromatography
RUO
29282, 65019, 9349
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P62829, P62830, P62832
Rpl23
RPL23 Fusion Protein Ag9120
20 μL
Primary
Rat, Mouse, Human