Learn More
RNASE11 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17203-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This antibody detected 30 kDa band with Testis and L02 cell lysate.
RNASE11 (Ribonuclease A Family Member 11 (Inactive)) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and ribonuclease activity.Spécification
RNASE11 | |
Polyclonal | |
Unconjugated | |
RNASE11 | |
C14orf6, Probable ribonuclease 11, RNase 11, RNASE11 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122651 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8TAA1 | |
RNASE11 | |
RNASE11 Fusion Protein Ag10981 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |