Learn More
RASSF6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 11921-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
RASSF6 is a member of the RAS association domain family of RAS effectors/tumor suppressors that mediate some of the growth inhibitory properties of RAS oncogenes.Spécification
RASSF6 | |
Polyclonal | |
Unconjugated | |
RASSF6 | |
RASSF6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
166824, 305251, 73246 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.53 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q4QR82, Q6ZTQ3, Q80UQ2 | |
RASSF6 | |
RASSF6 Fusion Protein Ag2565 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |