missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RALB Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 12340-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes a GTP-binding protein that belongs to the small GTPase superfamily and Ras family of proteins. GTP-binding proteins mediate the transmembrane signaling initiated by the occupancy of certain cell surface receptors.Spécification
RALB | |
Polyclonal | |
Unconjugated | |
RALB | |
RALB, Ras related protein Ral B | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5899, 64143 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P11234, Q9JIW9 | |
RALB | |
RALB Fusion Protein Ag2992 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |