Learn More
PSMG2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 10959-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Chaperone protein which promotes assembly of the 20S proteasome as part of a heterodimer with PSMG1. The PSMG1-PSMG2 heterodimer binds to the PSMA5 and PSMA7 proteasome subunits, promotes assembly of the proteasome alpha subunits into the heteroheptameric alpha ring and prevents alpha ring dimerization.Spécification
PSMG2 | |
Polyclonal | |
Unconjugated | |
PSMG2 | |
CLAST3, HCCA3, HsT1707, MDS003, PAC 2, PAC2, PSMG2, TNFSF5IP1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
107047, 291539, 56984 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q969U7, Q9EST4 | |
Psmg2 | |
PSMG2 Fusion Protein Ag1400 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |