missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 15549-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
This gene encodes an enzyme that catalyzes the phosphoribosylation of ribose 5-phosphate to 5-phosphoribosyl-1-pyrophosphate, which is necessary for purine metabolism and nucleotide biosynthesis. Defects in this gene are a cause of phosphoribosylpyrophosphate synthetase superactivity, Charcot-Marie-Tooth disease X-linked recessive type 5 and Arts Syndrome.Spécification
PRPS1 | |
Polyclonal | |
Unconjugated | |
PRPS1 | |
ARTS, CMTX5, KIAA0967, PPRibP, PRPS1, PRS I, PRSI | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
19139, 29562, 5631 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P60891, P60892, Q9D7G0 | |
Prps1 | |
PRPS1 Fusion Protein Ag7907 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |