Learn More
PPIL6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 17452-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
PPIases accelerate the folding of proteins. It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides.Spécification
PPIL6 | |
Polyclonal | |
Unconjugated | |
PPIL6 | |
Cyclophilin like protein PPIL6, PPIase, PPIL6, Rotamase PPIL6 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
285755, 685567, 73075 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8IXY8, Q9D6D8 | |
PPIL6 | |
PPIL6 Fusion Protein Ag11578 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |