missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PPAR gamma Mouse anti-Human, Clone: 4E12F10, Proteintech
Mouse Monoclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | PPAR gamma |
---|---|
Clone | 4E12F10 |
Concentration | 1 mg/mL |
Applications | ELISA, Flow Cytometry, Western Blot |
Classification | Monoclonal |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16847095
|
Proteintech
60127-1-IG-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16837095
|
Proteintech
60127-1-IG-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
PPP1R10 is a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene encodes a protein with similarity to a rat protein that has an inhibitory effect on protein phosphatase-1 (PP1). The rat protein localizes to the nucleus and colocalizes with chromatin at distinct phases during mitosis. This gene lies within the major histocompatibility complex class I region on chromosome 6.Spécification
PPAR gamma | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
CIMT1, NR1C3, PPAR gamma, PPARG, PPARG1 | |
PPARG | |
IgG1 | |
Caprylic Acid Precipitation | |
Antibody |
4E12F10 | |
ELISA, Flow Cytometry, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
P37231 | |
5468 | |
PPAR gamma Fusion Protein Ag10005 | |
Primary | |
-20°C | |
PPARG |