Learn More
Liebherr™ Positive Cabinet with White Epoxy Body
Positive Cabinet with White Epoxy Body is a fridge constructed of a white epoxy steel body with anti-corrosion treatment
Specifications
Gamme de températures | 2°C à 15°C |
---|---|
Réfrigérant | R-600a |
Matériau de la hotte | Epoxy |
Intérieur | Polystyrène |
Affichage | Numérique sur la porte |
Description
- Fully automatic defrost
- Mechanical thermostat
- Digital temperature display on door
- Closed door secure controls
- Easyclean tank thermoformed in food grade PS
- White epoxy steel body with anti-corrosion treatment
- High density 50 mm thick insulation without HFC
- Fan stops when the door is opened
- Reversible door with lock and automatic recall
- LED lighting with independent switch positioned on the ceiling
- Plastic coated steel grilles, max load 45 kg, removable with the door open at 90°
- Easily replaceable clip-on magnetic door seal
- R-600a clean fluid
Specifications
2°C à 15°C | |
Epoxy | |
Numérique sur la porte | |
Automatique | |
230 V |
R-600a | |
Polystyrène | |
Statique | |
2 A |