missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PON1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Spécification
Antigène | PON1 |
---|---|
Concentration | 0.15 mg/mL |
Applications | Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16818214
|
Proteintech
18155-1-AP-20UL |
20 μL |
145.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16808214
|
Proteintech
18155-1-AP-150UL |
150 μL |
410.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
Glycosylated PON1 migrates at 42-45 kDa. 55 kDa is a dimmer of isoform CRA-b. The antibody has cross-reaction to PON2 and PON3.
The enzyme encoded by this gene is an arylesterase that mainly hydrolyzes paroxon to produce p-nitrophenol. Paroxon is an organophosphorus anticholinesterase compound that is produced in vivo by oxidation of the insecticide parathion. Polymorphisms in this gene are a risk factor in coronary artery disease. The gene is found in a cluster of three related paraoxonase genes at 7q21.3.Spécification
PON1 | |
Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P27169, P52430, P55159, Q15165, Q15166, Q62086, Q62087, Q68FP2, Q6AXM8 | |
18979, 269823, 296851, 312086, 330260, 5444, 5445, 5446, 84024 | |
PON1 Fusion Protein Ag12934 | |
Primary | |
-20°C | |
PON1 |
0.15 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A esterase 1, Aromatic esterase 1, ESA, K 45, paraoxonase 1, PON, PON 1, PON1, Serum aryldialkylphosphatase 1 | |
Pon1, Pon2, PON3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |