Learn More
PLVAP Mouse anti-Human, Unconjugated, Clone: 1D4A6, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 67158-1-IG-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The PLVAP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PLVAP. This antibody recognizes human, pig antigen. The PLVAP antibody has been validated for the following applications: WB, ELISA analysis.
Spécification
PLVAP | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Pig |
Western Blot, ELISA | |
1D4A6 | |
Unconjugated | |
20 μL | |
83483 | |
IgG2a |