Learn More
Fluorure de perfluoro(2-méthyl-3-oxahexanoyle, 97 %, Thermo Scientific Chemicals
19.30€ - 180.00€
Identifiants chimiques
CAS | 2062-98-8 |
---|---|
Formule moléculaire | C6F12O2 |
Poids moléculaire (g/mol) | 332.045 |
Numéro MDL | MFCD00054657 |
Clé InChI | BCLQALQSEBVVAD-UHFFFAOYSA-N |
Synonyme | 2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd |
CID PubChem | 102740 |
Nom IUPAC | Fluorure de 2,3,3,3-tétrafluoro-2-1,1,2,2,3,3,3(-heptafluoropropoxy)propanoyl |
SMILES | C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
11389185
|
Thermo Scientific Alfa Aesar
L16483.03 |
1 g | |||||||
11399185
|
Thermo Scientific Alfa Aesar
L16483.06 |
5 g |
59.70€
5g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
11309195
|
Thermo Scientific Alfa Aesar
L16483.14 |
25 g |
180.00€
25g |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Identifiants chimiques
2062-98-8 | |
332.045 | |
BCLQALQSEBVVAD-UHFFFAOYSA-N | |
102740 | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F |
C6F12O2 | |
MFCD00054657 | |
2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd | |
Fluorure de 2,3,3,3-tétrafluoro-2-1,1,2,2,3,3,3(-heptafluoropropoxy)propanoyl |
Spécification
Perfluoro(2-methyl-3-oxahexanoyl) fluoride | |
54°C to 56°C | |
C6F12O2 | |
MFCD00054657 | |
Moisture sensitive | |
BCLQALQSEBVVAD-UHFFFAOYSA-N | |
Fluorure de 2,3,3,3-tétrafluoro-2-1,1,2,2,3,3,3(-heptafluoropropoxy)propanoyl | |
102740 | |
97% |
2062-98-8 | |
None | |
1.3 | |
UN3265 | |
2,3,3,3-tetrafluoro-2-perfluoropropoxy propanoyl fluoride, undecafluoro-2-methyl-3-oxahexanoyl fluoride, perfluoro 2-methyl-3-oxahexanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy propanoyl fluoride, perfluoro-2-methyl-3-oxahexanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-heptafluoropropoxy, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy propanoyl fluoride, propanoyl fluoride, 2,3,3,3-tetrafluoro-2-1,1,2,2,3,3,3-heptafluoropropoxy, hfpo dimer, acmc-1cljd | |
C(=O)(C(C(F)(F)F)(OC(C(C(F)(F)F)(F)F)(F)F)F)F | |
332.045 | |
332.06 |
Sécurité et traitement
GHS H Statement
H314-H318-H335
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 218-173-8
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only