missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NTHL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14918-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
Oxygen radicals damage chromosonal DNA causing cell death and inducing mutations. Although a major focus of oxidatively damaged DNA has centered on the repair of 8-oxo-G, a number of other damaged DNA sites are created by free-radical attack on DNA. The human NTH1 repair protein is one enzyme that has been shown to act on a large number of these other oxidatively damaged DNA sites.A homologue of E. coli Endonuclease III, NTH is a DNA glycosylase with apurinic/apyrimidinic lyase activity. The NTH protein has broad substrate specificity, including numerous ring saturation and fragmentation products of pyrimidines. Research indicates NTH plays a crucial role in removal of oxidative base lesions in mitochondrial DNA.Spécification
NTHL1 | |
Polyclonal | |
Unconjugated | |
NTHL1 | |
NTH1, NTHL1, OCTS3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
18207, 29541, 4913 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O35980, P78549 | |
Nthl1 | |
NTHL1 Fusion Protein Ag6734 | |
20 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |