Learn More
NOP14 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque:  Proteintech 26854-1-AP-20UL
 Afficher plus de versions de ce produit
Code produit. 16845195
  • 145.00€ / 20µL
Découvrez plus d'offres spéciales
Description
Description
Involved in nucleolar processing of pre-18S ribosomal RNA. Has a role in the nuclear export of 40S pre-ribosomal subunit to the cytoplasm.
Spécification
Spécification
NOP14
Polyclonal
Unconjugated
NOP14
C4orf9, NOL14, NOP14, Nucleolar complex protein 14, Nucleolar protein 14, RES4 25, RES425
Rabbit
Antigen Affinity Chromatography
RUO
289724, 75416, 8602
-20°C
Liquid
Western Blot, Immunocytochemistry, Immunofluorescence
0.37 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P78316, Q8R3N1
Nop14
NOP14 Fusion Protein Ag25476
20 μL
Primary
Mouse, Human, Rat