missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NFAM1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 14310-1-AP-20UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The protein encoded by this gene is a type I membrane receptor that activates cytokine gene promoters such as the IL-13 and TNF-alpha promoters. The encoded protein contains an immunoreceptor tyrosine-based activation motif (ITAM) and is thought to regulate the signaling and development of B-cells. It is also known CNAIP.Spécification
NFAM1 | |
Polyclonal | |
Unconjugated | |
NFAM1 | |
bK126B4.4, CNAIP, NFAM1, NFAT activation molecule 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
150372 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.17 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q8NET5 | |
NFAM1 | |
NFAM1 Fusion Protein Ag5505 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |