Learn More
NeutraControl PDGF-BB Mouse anti-Human, Unconjugated, Clone: 4E2C7, Proteintech
Mouse Monoclonal Antibody
Marque: Proteintech 69520-1-IG-100UG
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
The NeutraControl PDGF-BB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human NeutraControl PDGF-BB. This antibody recognizes human antigen. The NeutraControl PDGF-BB antibody has been validated for the following applications: Non-Neutralization, ELISA analysis.
Spécification
NeutraControl PDGF-BB | |
4E2C7 | |
Mouse | |
Primary | |
Human |
Monoclonal | |
Unconjugated | |
100 μg | |
5155 | |
IgG2a |