missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDFIP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
140.00€ - 399.00€
Spécification
Antigène | NDFIP1 |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugué | Unconjugated |
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Quantité | Prix | Quantité et disponibilité | |||||
16854844
|
Proteintech
15602-1-AP-20UL |
20 μL |
140.00€
20µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
16844844
|
Proteintech
15602-1-AP-150UL |
150 μL |
399.00€
150µL |
Veuillez vous connecter pour pouvoir commander cet article. Besoin d'un compte web? Créer le vôtre dès maintenant! | |||||
Description
The NEDD4 family-interacting protein 1 (NDFIP1) belongs to a small group of evolutionarily conserved proteins with three transmembrane domains and is an integral Golgi membrane protein. It is a potential target for ubiquitination by the Nedd4 family of proteins. NDFIP1 is strongly expressed in surviving neurons following acute cortical brain injury, and overexpression in cultured cortical neurons increased survival following growth factor starvation, suggesting that NDFIP1 may play a role in neuronal survival. NDFIP1 and the related protein NDFIP2 are thought to interact with and regulate multiple components of the EGF and PTEN/Akt signaling pathways. Recent studies suggest that NDFIP1 may also play a role in Th17 differentiation by limiting the production of proinflammatory cytokines.Spécification
NDFIP1 | |
Immunofluorescence, Immunocytochemistry, Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5U2S1, Q8R0W6, Q9BT67 | |
291609, 65113, 80762 | |
NDFIP1 Fusion Protein Ag7985 | |
Primary | |
-20°C | |
NDFIP1 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
N4WBP5, NDFIP1, PSEC0223 | |
Ndfip1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |