missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYCT1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marque: Proteintech 22004-1-AP-150UL
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
MYCT1 was initially identified as a novel target of the c-Myc oncogene in myeloid cells. It is a widely expressed nuclear protein whose overexpression can promote apoptosis, alteration of morphology, enhancement of anchorage-independent cell growth, tumorigenic conversion, promotion of genomic instability, and inhibition of hematopoietic differentiation. MYCT1 binds to the promoters of several c-Myc-regulated genes and it has been suggested that the phenotypes seen in MYCT1-overexpressing cells are a result of the deregulation of these genes. RUNX1-ETO, a fusion protein made up of RUNX1 and ETO, is thought to deregulate the proliferation and responsiveness of human hematopoietic progenitor cells downstream of MYCT1.Spécification
MYCT1 | |
Polyclonal | |
Unconjugated | |
MYCT1 | |
MTLC, MTMC1, myc target 1, Myc target protein 1, MYCT1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
68632, 80177 | |
-20°C | |
Liquid |
Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8N699, Q8R411 | |
Myct1 | |
MYCT1 Fusion Protein Ag4800 | |
150 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |